siRNA Details
| Gene Symbol: Mss51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029104.1 | NP_083380.1 | 7 | 1549 | ||
| GenBank mRNA | AK014794.1 | - | 7 | 1330 | ||
| BC119773.1 | - | 7 | 1442 | 40107223 | ||
| BC119816.1 | - | 7 | 1442 | 40107221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mss51| Gene Symbol: Mss51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029104.1 | NP_083380.1 | 7 | 1549 | ||
| GenBank mRNA | AK014794.1 | - | 7 | 1330 | ||
| BC119773.1 | - | 7 | 1442 | 40107223 | ||
| BC119816.1 | - | 7 | 1442 | 40107221 | ||