siRNA Details
| Gene Symbol: 4930447A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029113.1 | NP_083389.1 | 2, 3 | 317 | ||
| GenBank mRNA | AK015396.1 | - | 2, 3 | 319 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930447A16Rik| Gene Symbol: 4930447A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029113.1 | NP_083389.1 | 2, 3 | 317 | ||
| GenBank mRNA | AK015396.1 | - | 2, 3 | 319 | ||