siRNA Details
| Gene Symbol: Armc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081393.1 | NP_001074862.1 | 20 | 3105 | ||
| GenBank mRNA | BC158097.1 | - | 19 | 3094 | 9088178 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc4| Gene Symbol: Armc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081393.1 | NP_001074862.1 | 20 | 3105 | ||
| GenBank mRNA | BC158097.1 | - | 19 | 3094 | 9088178 | |