siRNA Details
| Gene Symbol: Csmd2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029137.1 | NP_083413.1 | 4 | 368 | ||
| GenBank mRNA | AK015502.1 | - | 4 | 369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csmd2os| Gene Symbol: Csmd2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029137.1 | NP_083413.1 | 4 | 368 | ||
| GenBank mRNA | AK015502.1 | - | 4 | 369 | ||