siRNA Details
| Gene Symbol: 4930480K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015602.1 | - | 2 | 903 | ||
| AK133329.1 | - | 2 | 575 | |||
| CJ047672.1 | - | 2 | 575 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930480K23Rik| Gene Symbol: 4930480K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015602.1 | - | 2 | 903 | ||
| AK133329.1 | - | 2 | 575 | |||
| CJ047672.1 | - | 2 | 575 | |||