siRNA Details
| Gene Symbol: Zcchc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029158.2 | NP_083434.1 | 1 | 267 | ||
| GenBank mRNA | AK015789.1 | - | 1 | 269 | ||
| BC145772.1 | - | 1 | 256 | 40131075 | ||
| BC145774.1 | - | 1 | 256 | 40131077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc13| Gene Symbol: Zcchc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029158.2 | NP_083434.1 | 1 | 267 | ||
| GenBank mRNA | AK015789.1 | - | 1 | 269 | ||
| BC145772.1 | - | 1 | 256 | 40131075 | ||
| BC145774.1 | - | 1 | 256 | 40131077 | ||