siRNA Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029162.2 | NP_083438.1 | 3 | 1115 | ||
| GenBank mRNA | AK015819.1 | - | 1 | 26 | ||
| AK043044.1 | - | 3 | 1110 | |||
| AK044991.1 | - | 3 | 1142 | |||
| AK140546.1 | - | 3 | 1110 | |||
| BC028988.1 | - | 3 | 1099 | 5376301 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK148068.1 | - | 1121 | Chr. - - [-] on Build | ||

Zbtb49
Genomic Map