siRNA Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029162.2 | NP_083438.1 | 6, 7 | 1563 | ||
| GenBank mRNA | AK015819.1 | - | 4, 5 | 475 | ||
| AK043044.1 | - | 6, 7 | 1558 | |||
| AK054488.1 | - | 3, 4 | 255 | |||
| AK140546.1 | - | 6, 7 | 1558 | |||
| BC028988.1 | - | 5, 6 | 1500 | 5376301 | ||
| BC029122.1 | - | 4, 5 | 376 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK148068.1 | - | 1569 | Chr. - - [-] on Build | ||

Zbtb49
Genomic Map