siRNA Details
Gene Symbol: 4930511M06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015760.1 | - | 2, 3 | 296 | ||
AK144587.1 | - | 2, 3 | 421 | |||
AK165876.1 | - | 2, 3 | 454 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930511M06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015760.1 | - | 2, 3 | 296 | ||
AK144587.1 | - | 2, 3 | 421 | |||
AK165876.1 | - | 2, 3 | 454 |