siRNA Details
| Gene Symbol: 4930511M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015760.1 | - | 3 | 363 | ||
| AK144587.1 | - | 3 | 488 | |||
| AK144776.1 | - | 2 | 366 | |||
| AK165835.1 | - | 172 | ||||
| AK165876.1 | - | 3 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930511M06Rik| Gene Symbol: 4930511M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015760.1 | - | 3 | 363 | ||
| AK144587.1 | - | 3 | 488 | |||
| AK144776.1 | - | 2 | 366 | |||
| AK165835.1 | - | 172 | ||||
| AK165876.1 | - | 3 | 521 | |||