siRNA Details
Gene Symbol: Gm9733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076679.2 | NP_001070147.1 | 2 | 395 | ||
GenBank mRNA | DQ055452.1 | - | 2 | 396 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm9733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076679.2 | NP_001070147.1 | 2 | 395 | ||
GenBank mRNA | DQ055452.1 | - | 2 | 396 |