siRNA Details
| Gene Symbol: Sv2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029210.1 | NP_083486.1 | 6, 7 | 1358 | ||
| GenBank mRNA | AK137053.1 | - | 2, 3 | 200 | ||
| AK173092.1 | - | 6, 7 | 1358 | |||
| BC132111.1 | - | 5, 6 | 1194 | 40130388 | ||
| BC137861.1 | - | 5, 6 | 1194 | 8860880 | ||

Sv2c
Genomic Map