siRNA Details
| Gene Symbol: Sv2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029210.1 | NP_083486.1 | 10 | 1811 | ||
| GenBank mRNA | AK015921.1 | - | 3 | 316 | ||
| AK137053.1 | - | 6 | 653 | |||
| AK173092.1 | - | 10 | 1811 | |||
| BC132111.1 | - | 9 | 1647 | 40130388 | ||
| BC137861.1 | - | 9 | 1647 | 8860880 | ||

Sv2c
Genomic Map