siRNA Details
Gene Symbol: Sv2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029210.1 | NP_083486.1 | 13 | 2254 | ||
GenBank mRNA | AK015921.1 | - | 6 | 759 | ||
AK173092.1 | - | 13 | 2254 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sv2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029210.1 | NP_083486.1 | 13 | 2254 | ||
GenBank mRNA | AK015921.1 | - | 6 | 759 | ||
AK173092.1 | - | 13 | 2254 |