siRNA Details
| Gene Symbol: Prr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165892.1 | NP_001159364.1 | 3 | 450 | ||
| NM_001282012.1 | NP_001268941.1 | 2 | 351 | |||
| NM_001282013.1 | NP_001268942.1 | 4 | 356 | |||
| NM_001282024.1 | NP_001268953.1 | 3 | 450 | |||
| NM_001282025.1 | NP_001268954.1 | 4 | 356 | |||
| NM_145487.3 | NP_663462.2 | 2 | 379 | |||
| GenBank mRNA | AJ504719.1 | - | 2 | 1148 | ||
| AK016009.1 | - | 3 | 268 | |||
| AK132945.1 | - | 2 | 79 | |||
| AK218162.1 | - | 2 | 78 | |||
| BC016636.1 | - | 2 | 300 | 2650633 | ||
| BE947267.1 | - | 4 | 339 | |||
| BU148736.1 | - | 4 | 313 | |||
| BY774318.1 | - | 3 | 278 | |||
| CJ049424.1 | - | 4 | 356 | |||
| CJ059038.1 | - | 2 | 380 | |||

Prr3
Genomic Map