siRNA Details
| Gene Symbol: Rnf121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029211.2 | NP_083487.2 | 1, 2 | 65 | ||
| GenBank mRNA | AK078509.1 | - | 1, 2 | 26 | ||
| AK159260.1 | - | 1, 2 | 92 | |||
| AK168821.1 | - | 1, 2 | 78 | |||
| AK169089.1 | - | 1, 2 | 78 | |||
| BC022686.1 | - | 1, 2 | 65 | 4241984 | ||

Rnf121
Genomic Map