siRNA Details
| Gene Symbol: Rnf19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029219.1 | NP_083495.1 | 5 | 1206 | ||
| GenBank mRNA | AK089131.1 | - | 4 | 445 | ||
| BC117807.1 | - | 5 | 764 | |||
| BC139808.1 | - | 5 | 767 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC038660.1 | - | 131 | Chr. - - [-] on Build | ||

Rnf19b
Genomic Map