siRNA Details
| Gene Symbol: Rnf19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029219.1 | NP_083495.1 | 7 | 1445 | ||
| GenBank mRNA | AK015966.1 | - | 2 | 129 | ||
| BC052529.1 | - | 1 | 47 | |||
| BC117807.1 | - | 7 | 1003 | |||
| BC139808.1 | - | 7 | 1006 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf19b| Gene Symbol: Rnf19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029219.1 | NP_083495.1 | 7 | 1445 | ||
| GenBank mRNA | AK015966.1 | - | 2 | 129 | ||
| BC052529.1 | - | 1 | 47 | |||
| BC117807.1 | - | 7 | 1003 | |||
| BC139808.1 | - | 7 | 1006 | |||