siRNA Details
| Gene Symbol: 4930564D02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029228.1 | NP_083504.1 | 2 | 56 | ||
| GenBank mRNA | AK016218.1 | - | 2 | 57 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930564D02Rik| Gene Symbol: 4930564D02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029228.1 | NP_083504.1 | 2 | 56 | ||
| GenBank mRNA | AK016218.1 | - | 2 | 57 | ||