siRNA Details
| Gene Symbol: 4930564B18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029230.1 | NP_083506.1 | 4, 5 | 622 | ||
| GenBank mRNA | AK016216.1 | - | 4, 5 | 623 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930564B18Rik| Gene Symbol: 4930564B18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029230.1 | NP_083506.1 | 4, 5 | 622 | ||
| GenBank mRNA | AK016216.1 | - | 4, 5 | 623 | ||