siRNA Details
| Gene Symbol: 4930553J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016107.1 | - | 476 | |||
| BC062172.1 | - | 1 | 350 | |||
| BC080822.1 | - | 1 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930553J12Rik| Gene Symbol: 4930553J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016107.1 | - | 476 | |||
| BC062172.1 | - | 1 | 350 | |||
| BC080822.1 | - | 1 | 338 | |||