siRNA Details
| Gene Symbol: Nop14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029278.2 | NP_083554.2 | 1, 2 | 299 | ||
| GenBank mRNA | AK011648.1 | - | 1, 2 | 275 | ||
| AK078605.1 | - | 1, 2 | 285 | |||
| AK079609.1 | - | 1, 2 | 301 | |||
| AK148605.1 | - | 1, 2 | 258 | |||
| AK161728.1 | - | 1, 2 | 237 | |||
| BC024998.1 | - | 1, 2 | 220 | 5012070 | ||
| BC043043.1 | - | 1, 2 | 242 | 5701430 | ||

Nop14
Genomic Map