siRNA Details
Gene Symbol: 1700006E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029287.1 | NP_083563.1 | 4 | 443 | ||
GenBank mRNA | AK005669.1 | - | 4 | 447 | ||
BC100517.1 | - | 4 | 418 | |||
BC116819.1 | - | 4 | 439 | 40126138 | ||
BC116821.1 | - | 4 | 439 | 40126140 |
