siRNA Details
| Gene Symbol: Cypt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029289.1 | NP_083565.1 | 1 | 282 | ||
| GenBank mRNA | AK005586.1 | - | 1 | 283 | ||
| AM040460.1 | - | 1 | 66 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cypt12| Gene Symbol: Cypt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029289.1 | NP_083565.1 | 1 | 282 | ||
| GenBank mRNA | AK005586.1 | - | 1 | 283 | ||
| AM040460.1 | - | 1 | 66 | |||