siRNA Details
| Gene Symbol: 1700011I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029290.1 | NP_083566.1 | 6, 7 | 624 | ||
| GenBank mRNA | CF981605.1 | - | 379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700011I03Rik| Gene Symbol: 1700011I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029290.1 | NP_083566.1 | 6, 7 | 624 | ||
| GenBank mRNA | CF981605.1 | - | 379 | |||