siRNA Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029291.1 | NP_083567.1 | 13 | 1245 | ||
| GenBank mRNA | AK051387.1 | - | 13 | 1294 | ||
| AK148105.1 | - | 13 | 1277 | |||
| BC013537.1 | - | 13 | 1245 | 4240784 | ||
| BC024840.1 | - | 6 | 396 | |||

Ascc2
Genomic Map