siRNA Details
| Gene Symbol: Iqcf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029300.1 | NP_083576.1 | 2 | 270 | ||
| GenBank mRNA | AK005734.1 | - | 2 | 271 | ||
| BC125546.1 | - | 2 | 229 | 40130061 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcf5| Gene Symbol: Iqcf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029300.1 | NP_083576.1 | 2 | 270 | ||
| GenBank mRNA | AK005734.1 | - | 2 | 271 | ||
| BC125546.1 | - | 2 | 229 | 40130061 | ||