siRNA Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081275.1 | NP_001074744.1 | 4 | 417 | ||
| GenBank mRNA | AK005804.1 | - | 4 | 419 | ||
| BC061017.1 | - | 3 | 143 | 6774925 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap126| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081275.1 | NP_001074744.1 | 4 | 417 | ||
| GenBank mRNA | AK005804.1 | - | 4 | 419 | ||
| BC061017.1 | - | 3 | 143 | 6774925 | ||