siRNA Details
| Gene Symbol: Pfn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029303.2 | NP_083579.1 | 1 | 285 | ||
| GenBank mRNA | AK005930.1 | - | 1 | 277 | ||
| BC048629.1 | - | 1 | 285 | 6744051 | ||
| Gene Symbol: BC038268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038268.1 | - | 1 | 53 | ||

Pfn3
Genomic Map