siRNA Details
| Gene Symbol: Hspb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029307.1 | NP_083583.1 | 1 | 107 | ||
| GenBank mRNA | AK005714.1 | - | 1 | 30 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspb9| Gene Symbol: Hspb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029307.1 | NP_083583.1 | 1 | 107 | ||
| GenBank mRNA | AK005714.1 | - | 1 | 30 | ||