siRNA Details
| Gene Symbol: 1700010B08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029308.1 | NP_083584.1 | 3 | 732 | ||
| GenBank mRNA | AK005811.1 | - | 3 | 693 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700010B08Rik| Gene Symbol: 1700010B08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029308.1 | NP_083584.1 | 3 | 732 | ||
| GenBank mRNA | AK005811.1 | - | 3 | 693 | ||