siRNA Details
| Gene Symbol: 1700013F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029314.1 | NP_083590.1 | 3 | 258 | ||
| GenBank mRNA | AK005946.1 | - | 3 | 277 | ||
| BC049763.1 | - | 3 | 296 | |||
| BC100493.1 | - | 291 | ||||
| BC116222.1 | - | 3 | 259 | |||
| BC116223.1 | - | 3 | 258 | 40091335 | ||

1700013F07Rik
Genomic Map