siRNA Details
Gene Symbol: Izumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029317.1 | NP_083593.1 | 5 | 464 | ||
GenBank mRNA | AK006268.1 | - | 5 | 438 | ||
AV260738.1 | - | 1 | 76 | |||
BC048526.1 | - | 6 | 501 | 6772588 |
