siRNA Details
Gene Symbol: 1700018C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029324.2 | NP_083600.1 | 3 | 174 | ||
GenBank mRNA | AK006081.1 | - | 3 | 175 | ||
BC048618.1 | - | 3 | 177 | 6742642 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700018C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029324.2 | NP_083600.1 | 3 | 174 | ||
GenBank mRNA | AK006081.1 | - | 3 | 175 | ||
BC048618.1 | - | 3 | 177 | 6742642 |