siRNA Details
| Gene Symbol: 1700018C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029324.2 | NP_083600.1 | 4, 5 | 356 | ||
| GenBank mRNA | AK006081.1 | - | 4, 5 | 357 | ||
| BC048618.1 | - | 4, 5 | 359 | 6742642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700018C11Rik| Gene Symbol: 1700018C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029324.2 | NP_083600.1 | 4, 5 | 356 | ||
| GenBank mRNA | AK006081.1 | - | 4, 5 | 357 | ||
| BC048618.1 | - | 4, 5 | 359 | 6742642 | ||