siRNA Details
| Gene Symbol: 1700015G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195601.1 | NP_001182530.1 | 2, 3 | 119 | ||
| GenBank mRNA | AK005994.1 | - | 2, 3 | 122 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700015G11Rik| Gene Symbol: 1700015G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195601.1 | NP_001182530.1 | 2, 3 | 119 | ||
| GenBank mRNA | AK005994.1 | - | 2, 3 | 122 | ||