siRNA Details
| Gene Symbol: Fam71e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028169.1 | NP_082445.1 | 4 | 453 | ||
| GenBank mRNA | AB473729.1 | - | 4 | 353 | ||
| AK006222.1 | - | 3 | 584 | |||
| AK006229.1 | - | 3 | 571 | |||
| BC049752.1 | - | 4 | 401 | 6773108 | ||
| BC100470.1 | - | 4 | 404 | |||
| BC130017.1 | - | 4 | 396 | 40140477 | ||

Fam71e1
Genomic Map