siRNA Details
| Gene Symbol: Nhej1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029342.4 | NP_083618.3 | 270 | |||
| GenBank mRNA | AK006481.1 | - | 264 | |||
| BC006063.1 | - | 244 | 3591824 | |||
| BC107252.1 | - | 189 | 40052353 | |||
| BC107253.1 | - | 189 | 40052354 | |||
| BC132359.1 | - | 160 | 40130636 | |||
| BC132361.1 | - | 160 | 40130638 | |||
| BY095373.1 | - | 193 | ||||

Nhej1
Genomic Map