siRNA Details
| Gene Symbol: Ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127346.1 | NP_001120818.1 | 4 | 393 | ||
| GenBank mRNA | AB183434.1 | - | 4 | 337 | ||
| AK007894.1 | - | 4 | 384 | |||
| AK166492.1 | - | 4 | 360 | |||
| AW123826.1 | - | 1 | 300 | |||
| AY262012.1 | - | 4 | 365 | |||
| BC116975.1 | - | 4 | 359 | 40126294 | ||
| BC116977.1 | - | 4 | 359 | 40126296 | ||
| BC144716.1 | - | 3 | 318 | |||
| BY251841.1 | - | 4 | 394 | |||

Ndufaf2
Genomic Map