siRNA Details
| Gene Symbol: Tm4sf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029360.3 | NP_083636.2 | 4 | 516 | ||
| GenBank mRNA | AI323987.1 | - | 2 | 463 | ||
| AK008064.1 | - | 4 | 514 | |||
| BC010782.1 | - | 4 | 476 | 4219239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm4sf5| Gene Symbol: Tm4sf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029360.3 | NP_083636.2 | 4 | 516 | ||
| GenBank mRNA | AI323987.1 | - | 2 | 463 | ||
| AK008064.1 | - | 4 | 514 | |||
| BC010782.1 | - | 4 | 476 | 4219239 | ||