siRNA Details
| Gene Symbol: Wnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290311.1 | NP_001277240.1 | 21 | 5319 | ||
| NM_001290313.1 | NP_001277242.1 | 21 | 5319 | |||
| NM_029361.4 | NP_083637.2 | 21 | 5319 | |||
| GenBank mRNA | AK044381.1 | - | 2 | 236 | ||
| AK129439.1 | - | 10 | 2176 | |||
| AK147550.1 | - | 21 | 5006 | |||
| BC046464.1 | - | 14 | 3118 | |||
| BC055795.1 | - | 2 | 214 | |||
| BC060187.1 | - | 10 | 2350 | |||

Wnk2
Genomic Map