siRNA Details
| Gene Symbol: Tex30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029368.1 | NP_083644.1 | 5 | 440 | ||
| GenBank mRNA | AK006487.1 | - | 3 | 472 | ||
| AK160565.1 | - | 5 | 441 | |||
| BC099557.1 | - | 4 | 353 | 30916402 | ||
| BC116881.1 | - | 3 | 637 | 40126200 | ||
| BC119032.1 | - | 3 | 705 | 8733781 | ||

Tex30
Genomic Map