siRNA Details
Gene Symbol: Spata25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029370.1 | NP_083646.1 | 2 | 155 | ||
GenBank mRNA | AK006145.1 | - | 2 | 130 | ||
BC116354.1 | - | 1 | 487 | 40099912 | ||
BC116355.1 | - | 2 | 117 | 40099919 |
