siRNA Details
| Gene Symbol: Spata25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029370.1 | NP_083646.1 | 2 | 712 | ||
| GenBank mRNA | AK006145.1 | - | 2 | 687 | ||
| BC116354.1 | - | 1 | 1044 | 40099912 | ||
| BC116355.1 | - | 2 | 674 | 40099919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata25| Gene Symbol: Spata25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029370.1 | NP_083646.1 | 2 | 712 | ||
| GenBank mRNA | AK006145.1 | - | 2 | 687 | ||
| BC116354.1 | - | 1 | 1044 | 40099912 | ||
| BC116355.1 | - | 2 | 674 | 40099919 | ||