siRNA Details
| Gene Symbol: Iqcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029408.2 | NP_083684.1 | 4 | 1147 | ||
| GenBank mRNA | AK017032.1 | - | 4 | 1147 | ||
| BC125429.1 | - | 4 | 1141 | 40129944 | ||
| BC137644.1 | - | 4 | 1141 | 8860663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcd| Gene Symbol: Iqcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029408.2 | NP_083684.1 | 4 | 1147 | ||
| GenBank mRNA | AK017032.1 | - | 4 | 1147 | ||
| BC125429.1 | - | 4 | 1141 | 40129944 | ||
| BC137644.1 | - | 4 | 1141 | 8860663 | ||