siRNA Details
| Gene Symbol: Gm15545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087248.1 | - | 2 | 903 | ||
| Gene Symbol: Bcl2l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029410.3 | NP_083686.1 | 3, 4 | 385 | ||
| GenBank mRNA | AK013411.1 | - | 3, 4 | 359 | ||
| AK017362.1 | - | 3, 4 | 386 | |||
| BC039807.1 | - | 537 | 1529147 | |||

Gm15545
Genomic Map