siRNA Details
| Gene Symbol: Bcl2l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029410.3 | NP_083686.1 | 6 | 569 | ||
| GenBank mRNA | AK017362.1 | - | 6 | 570 | ||
| BC039807.1 | - | 1090 | 1529147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl2l12| Gene Symbol: Bcl2l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029410.3 | NP_083686.1 | 6 | 569 | ||
| GenBank mRNA | AK017362.1 | - | 6 | 570 | ||
| BC039807.1 | - | 1090 | 1529147 | |||