siRNA Details
| Gene Symbol: 4930402K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270700.1 | NP_001257629.1 | 1 | 900 | ||
| GenBank mRNA | AK015052.1 | - | 1 | 901 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930402K13Rik| Gene Symbol: 4930402K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270700.1 | NP_001257629.1 | 1 | 900 | ||
| GenBank mRNA | AK015052.1 | - | 1 | 901 | ||