siRNA Details
| Gene Symbol: 1700007G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024614.1 | NP_001019785.1 | 4 | 395 | ||
| GenBank mRNA | AK005711.1 | - | 5 | 524 | ||
| BC049770.1 | - | 4 | 395 | 6774684 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700007G11Rik| Gene Symbol: 1700007G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024614.1 | NP_001019785.1 | 4 | 395 | ||
| GenBank mRNA | AK005711.1 | - | 5 | 524 | ||
| BC049770.1 | - | 4 | 395 | 6774684 | ||