siRNA Details
| Gene Symbol: Tex26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029464.1 | NP_083740.1 | 4 | 425 | ||
| GenBank mRNA | AK016369.1 | - | 5 | 422 | ||
| AK087319.1 | - | 4 | 431 | |||
| AK161244.1 | - | 5 | 424 | |||
| BC120819.1 | - | 4 | 425 | 40129742 | ||
| BC144997.1 | - | 3 | 340 | |||
| BC144998.1 | - | 3 | 340 | |||
| BC144999.1 | - | 4 | 422 | 9053547 | ||

Tex26
Genomic Map